Learn More
Biolegend PE anti-mouse Notch 1
Supplier:  Biolegend 130607
Encompass_Preferred
PE anti-mouse Notch 1 [HMN1-12]; Isotype: Armenian Hamster IgG; Reactivity: Mouse; Apps: FC; Size: 50 ug
Catalog No. 50-164-580