Learn More
Biolegend PE anti-mouse/rat CD61
Supplier:  Biolegend 104308
Encompass_Preferred
PE anti-mouse/rat CD61 [2C9.G2 (HMβ3-1)]; Isotype: Armenian Hamster IgG; Reactivity: Mouse, Rat; Apps: FC; Size: 200 ug
Catalog No. 50-604-835