Learn More
Biolegend PE anti-mouse TCR Vα11.1, 11.2
Supplier:  Biolegend 139903
Encompass_Preferred
PE anti-mouse TCR Vα11.1, 11.2 [RR8-1]; Isotype: Rat IgG2b, κ; Reactivity: Mouse; Apps: FC; Size: 25 ug
Catalog No. 50-165-106