Learn More
Biolegend PE anti-mouse TCR Vα8.3
Supplier:  Biolegend 125707
Encompass_Preferred
PE anti-mouse TCR Vα8.3 [KT50]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 100 ug
Catalog No. 50-164-269