Learn More
Biolegend PE anti-mouse TCR Vβ11
Supplier:  Biolegend 139004
Encompass_Preferred
PE anti-mouse TCR Vβ11 [RR3-15]; Isotype: Rat IgG2b, κ; Reactivity: Mouse; Apps: FC; Size: 100 ug
Catalog No. 50-165-067