Learn More
Biolegend PE anti-mouse TCR Vβ2
Supplier:  Biolegend 127907
Encompass_Preferred
PE anti-mouse TCR Vβ2 [B20.6]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 25 ug
Catalog No. 50-164-426