Learn More
Biolegend PE anti-mouse CD365 (Tim-1)
Supplier:  Biolegend 119505
Encompass_Preferred
PE anti-mouse CD365 (Tim-1) [RMT1-4]; Isotype: Rat IgG2b, κ; Reactivity: Mouse; Apps: FC; Size: 50 ug
Catalog No. NC0167655