Learn More
Biolegend PE anti-rat CD45RC [OX-22]; Isotype: Mouse IgG1, ; Reactivity: Rat; Apps: FC; Size: 50 ug
Supplier:  Biolegend 204007
Encompass_Preferred
PE anti-rat CD45RC [OX-22]; Isotype: Mouse IgG1, ; Reactivity: Rat; Apps: FC; Size: 50 ug
Catalog No. 50-165-444