Learn More
Biolegend PE anti-rat IFN-γ
Supplier:  Biolegend 507806
Encompass_Preferred
PE anti-rat IFN-γ [DB-1]; Isotype: Mouse IgG1, κ; Reactivity: Mouse, Rat; Apps: ICFC; Size: 100 tests
Catalog No. 50-170-040