Learn More
Biolegend PE anti-rat IL-4 [OX-81]; Isotype: Mouse IgG1, ; Reactivity: Rat; Apps: ICFC; Size: 25 ug
Supplier:  Biolegend 511905
Encompass_Preferred
PE anti-rat IL-4 [OX-81]; Isotype: Mouse IgG1, ; Reactivity: Rat; Apps: ICFC; Size: 25 ug
Catalog No. 50-170-109