Learn More
Biolegend PE anti-RPS6
Supplier:  Biolegend 691804
Encompass_Preferred
PE anti-RPS6 [A16009C]; Isotype: Mouse IgG2b, κ; Reactivity: Human, Mouse; Apps: ICFC; Size: 100 tests
Catalog No. 50-208-7605