Learn More
Biolegend PE anti-T-bet [4B10]; Isotype: Mouse IgG1, ; Reactivity: Human, Mouse; Apps: ICFC; Size: 100 ug
Supplier:  Biolegend 644810
Encompass_Preferred
PE anti-T-bet [4B10]; Isotype: Mouse IgG1, ; Reactivity: Human, Mouse; Apps: ICFC; Size: 100 ug
Catalog No. 50-170-980