Learn More
Biolegend PE/Cyanine7 anti-ERK1/2 Phospho (Thr202/Tyr204)
Supplier:  Biolegend 369515
Encompass_Preferred
PE/Cyanine7 anti-ERK1/2 Phospho (Thr202/Tyr204) [6B8B69]; Isotype: Mouse IgG2a, κ; Reactivity: Human, Mouse; Apps: ICFC; Size: 25 tests
Catalog No. 50-207-8513