Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC PE-conjugated Donkey Anti-Rabbit IgG H&L | 100 UL
SDP

Catalog No. 5000444120
Encompass_Preferred

PE-conjugated Donkey Anti-Rabbit IgG H&L is a polyclonal antibody, derived from Donkey, and conjugated with Phycoerythrin (PE). It specifically targets and binds to the light and heavy chains of Rabbit IgG antibodies, making it suitable for various immunological applications. This antibody is supplied in 0.01M Phosphate Buffered Saline, pH 7.2, containing 1% BSA, 50% glycerol, and 0.02% Sodium Azide.

  • Host: Donkey
  • Clonality: Polyclonal
  • Reactivity: Rabbit
  • Conjugated with Phycoerythrin (PE)
  • Affinity purified
  • Concentration: 1.0 mg/mL
  • Suitable for IHC-P, ICC/IF, FC, and ELISA applications
  • Formulated in 0.01M Phosphate Buffered Saline with BSA, glycerol, and sodium azide
  • Stable for 1 year when stored at -20°C, protected from light

Catalog No. 50-004-44120 Supplier Medchemexpress LLC Supplier No. HYP83745100UL
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

PE-conjugated Donkey Anti-Rabbit IgG H&L is a polyclonal antibody, derived from Donkey, and conjugated with Phycoerythrin (PE). It specifically targets and binds to the light and heavy chains of Rabbit IgG antibodies, making it suitable for various immunological applications. This antibody is supplied in 0.01M Phosphate Buffered Saline, pH 7.2, containing 1% BSA, 50% glycerol, and 0.02% Sodium Azide.

  • Host: Donkey
  • Clonality: Polyclonal
  • Reactivity: Rabbit
  • Conjugated with Phycoerythrin (PE)
  • Affinity purified
  • Concentration: 1.0 mg/mL
  • Suitable for IHC-P, ICC/IF, FC, and ELISA applications
  • Formulated in 0.01M Phosphate Buffered Saline with BSA, glycerol, and sodium azide
  • Stable for 1 year when stored at -20°C, protected from light

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.