Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC PE-conjugated Donkey Anti-Rabbit IgG H&L | 50 UL
SDP

Catalog No. 5000443358
Encompass_Preferred

This PE-conjugated antibody is a donkey originated polyclonal antibody that targets Goat IgG. It specifically binds to the light and heavy chains of rabbit IgG antibodies and is intended for research use only. The product is supplied as an affinity-purified liquid in 0.01M Phosphate Buffered Saline, pH 7.2, containing 1% BSA, 50% glycerol, and 0.02% Sodium Azide.

  • Conjugated with PE
  • Donkey originated polyclonal antibody
  • Targets Goat IgG antibody
  • Binds to light and heavy chains of rabbit IgG
  • For research use only
  • Affinity purified
  • Liquid appearance
  • Formulated in 0.01M PBS, pH 7.2, with 1% BSA, 50% glycerol, and 0.02% Sodium Azide
  • Store at -20°C for 1 year, protected from light
  • Avoid repeated freeze/thaw cycles
  • Shipped with blue ice
  • Suitable for IHC-P, ICC/IF, FC, and ELISA applications

Catalog No. 50-004-43358 Supplier Medchemexpress LLC Supplier No. HYP8374550UL
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This PE-conjugated antibody is a donkey originated polyclonal antibody that targets Goat IgG. It specifically binds to the light and heavy chains of rabbit IgG antibodies and is intended for research use only. The product is supplied as an affinity-purified liquid in 0.01M Phosphate Buffered Saline, pH 7.2, containing 1% BSA, 50% glycerol, and 0.02% Sodium Azide.

  • Conjugated with PE
  • Donkey originated polyclonal antibody
  • Targets Goat IgG antibody
  • Binds to light and heavy chains of rabbit IgG
  • For research use only
  • Affinity purified
  • Liquid appearance
  • Formulated in 0.01M PBS, pH 7.2, with 1% BSA, 50% glycerol, and 0.02% Sodium Azide
  • Store at -20°C for 1 year, protected from light
  • Avoid repeated freeze/thaw cycles
  • Shipped with blue ice
  • Suitable for IHC-P, ICC/IF, FC, and ELISA applications

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.