Learn More
MSC PIP CoreShield Engineered Yarn Glove, A4 Cut, 18G, ESD, 1/Pr.
Catalog No. 5000509792
Encompass
These lightweight 18 gauge seamless knit gloves provide A4 cut protection and are designed for comfort, dexterity, and tactile sensitivity in dry and slightly oily environments. Featuring a foam nitrile coating on the palm and fingers, they offer enhanced grip and are reinforced between the thumb and forefinger for durability. This ESD-safe, touchscreen-compatible glove is constructed from an HPPE and carbon fiber liner that is entirely fiberglass-free and food-certified.
  • Fiberglass-free and food-certified construction
  • Foam nitrile coating provides enhanced grip in dry and slightly oily conditions
  • Touchscreen compatible and ESD-safe
  • Reinforced thumb crotch for extended product life
  • Seamless knit wrist prevents dirt and debris from entering
  • Silicone-free and Oeko-Tex® standard 100 certified
  • ANSI level 3 abrasion and puncture resistance
Catalog No. 50-005-09792 Supplier MSC Supplier No. 71482657
These lightweight 18 gauge seamless knit gloves provide A4 cut protection and are designed for comfort, dexterity, and tactile sensitivity in dry and slightly oily environments. Featuring a foam nitrile coating on the palm and fingers, they offer enhanced grip and are reinforced between the thumb and forefinger for durability. This ESD-safe, touchscreen-compatible glove is constructed from an HPPE and carbon fiber liner that is entirely fiberglass-free and food-certified.
  • Fiberglass-free and food-certified construction
  • Foam nitrile coating provides enhanced grip in dry and slightly oily conditions
  • Touchscreen compatible and ESD-safe
  • Reinforced thumb crotch for extended product life
  • Seamless knit wrist prevents dirt and debris from entering
  • Silicone-free and Oeko-Tex® standard 100 certified
  • ANSI level 3 abrasion and puncture resistance