Learn More
Biolegend PE/Cyanine5 anti-human CD117 (c-kit)
Supplier:  Biolegend 323412
Encompass_Preferred
PE/Cyanine5 anti-human CD117 (c-kit) [A3C6E2]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 100 tests
Catalog No. 50-167-193