Learn More
Biolegend PE/Cyanine5 anti-human CD117 (c-kit)
Supplier:  Biolegend 313210
Encompass_Preferred
PE/Cyanine5 anti-human CD117 (c-kit) [104D2]; Isotype: Mouse IgG1, κ; Reactivity: Human, Cross-Reactivity: Cynomolgus, Cattle (Bovine, Cow); Apps: FC; Size: 100 tests
Catalog No. 50-166-724