Learn More
Biolegend PE/Cyanine5 anti-human CD123
Supplier:  Biolegend 306008
Encompass_Preferred
PE/Cyanine5 anti-human CD123 [6H6]; Isotype: Mouse IgG1, κ; Reactivity: Human, Cross-Reactivity: Rhesus; Apps: FC; Size: 100 tests
Catalog No. 50-166-313