Learn More
Biolegend PE/Cyanine5 anti-human CD18
Supplier:  Biolegend 302109
Encompass_Preferred
PE/Cyanine5 anti-human CD18 [TS1/18]; Isotype: Mouse IgG1, κ; Reactivity: Human, Cross-Reactivity: Swine (Pig, Porcine); Apps: FC; Size: 25 tests
Catalog No. 50-165-814