Learn More
Biolegend PE/Cyanine5 anti-human CD235ab
Supplier:  Biolegend 306606
Encompass_Preferred
PE/Cyanine5 anti-human CD235ab [HIR2]; Isotype: Mouse IgG2b, κ; Reactivity: Human, African Green, Baboon; Apps: FC; Size: 100 ug
Catalog No. 50-166-366