Learn More
Biolegend PE/Cyanine5 anti-human CD29
Supplier:  Biolegend 303005
Encompass_Preferred
PE/Cyanine5 anti-human CD29 [TS2/16]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green, Baboon, Cattle (Bovine, Cow), Cynomolgus, Dog (Canine), (Horse reactivity), Rhesus; Apps: FC; Size: 25 tests
Catalog No. 50-165-940