Learn More
Biolegend PE/Cyanine5 anti-human CD3
Supplier:  Biolegend 300309
Encompass_Preferred
PE/Cyanine5 anti-human CD3 [HIT3a]; Isotype: Mouse IgG2a, κ; Reactivity: Human; Apps: FC; Size: 25 tests
Catalog No. 50-165-534