Learn More
Biolegend PE/Cyanine5 anti-human CD33
Supplier:  Biolegend 303406
Encompass_Preferred
PE/Cyanine5 anti-human CD33 [WM53]; Isotype: Mouse IgG1, κ; Reactivity: Human, Chimpanzee; Apps: FC; Size: 100 tests
Catalog No. 50-165-985