Learn More
Biolegend PE/Cyanine5 anti-mouse CD140a
Supplier:  Biolegend 135919
Encompass_Preferred
PE/Cyanine5 anti-mouse CD140a [APA5]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 25 ug
Catalog No. 50-207-1157