Learn More
Biolegend PE/Cyanine5 anti-mouse CD4
Supplier:  Biolegend 130312
Encompass_Preferred
PE/Cyanine5 anti-mouse CD4 [H129.19]; Isotype: Rat IgG2a, κ; Reactivity: Mouse ; Apps: FC; Size: 100 ug
Catalog No. 50-164-565