Learn More
Biolegend PE/Cyanine5 anti-mouse CD48
Supplier:  Biolegend 103420
Encompass_Preferred
PE/Cyanine5 anti-mouse CD48 [HM48-1]; Isotype: Armenian Hamster IgG; Reactivity: Mouse; Apps: FC; Size: 100 ug
Catalog No. 50-162-845