Learn More
Biolegend PE/Cyanine5 anti-mouse CD80
Supplier:  Biolegend 104712
Encompass_Preferred
PE/Cyanine5 anti-mouse CD80 [16-10A1]; Isotype: Armenian Hamster IgG; Reactivity: Mouse, Cross-Reactivity: Dog (Canine); Apps: FC; Size: 100 ug
Catalog No. 50-162-973