Learn More
Biolegend PE/Cyanine5 anti-mouse CD86
Supplier:  Biolegend 105016
Encompass_Preferred
PE/Cyanine5 anti-mouse CD86 [GL-1]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 100 ug
Catalog No. 50-163-004