Learn More
Biolegend PE/Cyanine5 anti-mouse TCR β chain
Supplier:  Biolegend 109210
Encompass_Preferred
PE/Cyanine5 anti-mouse TCR β chain [H57-597]; Isotype: Armenian Hamster IgG; Reactivity: Mouse; Apps: FC; Size: 100 ug
Catalog No. 50-163-346