Learn More
Biolegend PE/Cyanine5 anti-mouse TER-119/Erythroid Cells
Supplier:  Biolegend 116210
Encompass_Preferred
PE/Cyanine5 anti-mouse TER-119/Erythroid Cells [TER-119]; Isotype: Rat IgG2b, κ; Reactivity: Mouse; Apps: FC; Size: 100 ug
Catalog No. 50-163-626