Learn More
Biolegend PE/Cyanine5 Mouse IgG1, κ isotype Ctrl
Supplier:  Biolegend 400118
Encompass_Preferred
PE/Cyanine5 Mouse IgG1, κ isotype Ctrl [MOPC-21]; Isotype: Mouse IgG1, κ; Reactivity: Mouse ; Apps: FC, ICFC; Size: 100 tests
Catalog No. 50-168-835