Learn More
Biolegend PE/Cyanine5 Mouse IgG2b, κ isotype Ctrl
Supplier:  Biolegend 400317
Encompass_Preferred
PE/Cyanine5 Mouse IgG2b, κ isotype Ctrl [MPC-11]; Isotype: Mouse IgG2b, κ; Reactivity: Mouse ; Apps: FC, ICFC; Size: 25 tests
Catalog No. 50-168-904