Learn More
Biolegend PE/Cyanine5 Rat IgG1, κ isotype Ctrl
Supplier:  Biolegend 400410
Encompass_Preferred
PE/Cyanine5 Rat IgG1, κ isotype Ctrl [RTK2071]; Isotype: Rat IgG1, κ; Apps: FC, ICFC; Size: 100 ug
Catalog No. 50-168-925