Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytek Biosciences Inc PE-Cy7 aHu CD19 Anti Primary

Supplier:  Cytek Biosciences Inc 600198T100

Encompass_Preferred

Product Name: PE-Cyanine7 Anti-Human CD19 (SJ25C1)
Product Class: Antibody
Type: Primary
Product Name: PE-Cyanine7 Anti-Human CD19 (SJ25C1)
Cytek Catalog Number: 60-0198-T100
Marker: CD19
Target Species: Human
Clonality: Monoclonal
Clone: SJ25C1
Host Species: Mouse
Isotype: IgG1, k
Conjugation: PE-Cyanine7
Size: 100 Tests

Catalog No. 50-000-87088


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.