Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytek Biosciences Inc PE-Cy7 aHu CD28 Anti Primary

Catalog No. 5000086792
Encompass_Preferred

Product Name: PE-Cyanine7 Anti-Human CD28 (CD28.2)
Product Class: Antibody
Type: Primary
Product Name: PE-Cyanine7 Anti-Human CD28 (CD28.2)
Cytek Catalog Number: 60-0289-T100
Marker: CD28
Target Species: Human
Reported Cross Reactivity: Baboon|Chimpanzee|Cynomolgus Monkey|Rhesus Macaque
Clonality: Monoclonal
Clone: CD28.2
Host Species: Mouse
Isotype: IgG1, k
Conjugation: PE-Cyanine7
Size: 100 Tests

Catalog No. 50-000-86792 Supplier Cytek Biosciences Inc Supplier No. 600289T100
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Product Name: PE-Cyanine7 Anti-Human CD28 (CD28.2)
Product Class: Antibody
Type: Primary
Product Name: PE-Cyanine7 Anti-Human CD28 (CD28.2)
Cytek Catalog Number: 60-0289-T100
Marker: CD28
Target Species: Human
Reported Cross Reactivity: Baboon|Chimpanzee|Cynomolgus Monkey|Rhesus Macaque
Clonality: Monoclonal
Clone: CD28.2
Host Species: Mouse
Isotype: IgG1, k
Conjugation: PE-Cyanine7
Size: 100 Tests

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.