Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytek Biosciences Inc PE-Cy7 aMu CD3e Anti Primary

Catalog No. 5000086791
Encompass_Preferred

Product Name: PE-Cyanine7 Anti-Mouse CD3e (145-2C11)
Product Class: Antibody
Type: Primary
Product Name: PE-Cyanine7 Anti-Mouse CD3e (145-2C11)
Cytek Catalog Number: 60-0031-U025
Marker: CD3e
Target Species: Mouse
Clonality: Monoclonal
Clone: 145-2C11
Host Species: Armenian Hamster
Isotype: IgG
Conjugation: PE-Cyanine7
Size: 25 μg

Catalog No. 50-000-86791 Supplier Cytek Biosciences Inc Supplier No. 600031U025
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Product Name: PE-Cyanine7 Anti-Mouse CD3e (145-2C11)
Product Class: Antibody
Type: Primary
Product Name: PE-Cyanine7 Anti-Mouse CD3e (145-2C11)
Cytek Catalog Number: 60-0031-U025
Marker: CD3e
Target Species: Mouse
Clonality: Monoclonal
Clone: 145-2C11
Host Species: Armenian Hamster
Isotype: IgG
Conjugation: PE-Cyanine7
Size: 25 μg

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.