Learn More
Biolegend PE/Cyanine7 anti-human CD158e1 (KIR3DL1, NKB1)
Supplier:  Biolegend 312720
Encompass_Preferred
PE/Cyanine7 anti-human CD158e1 (KIR3DL1, NKB1) [DX9]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 100 tests
Catalog No. 50-712-016