Learn More
Biolegend PE/Cyanine7 anti-human CD11b (activated)
Supplier:  Biolegend 301412
Encompass_Preferred
PE/Cyanine7 anti-human CD11b (activated) [CBRM1/5]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 100 tests
Catalog No. 50-711-974