Learn More
Biolegend PE/Cyanine7 anti-human CD120b
Supplier:  Biolegend 358412
Encompass_Preferred
PE/Cyanine7 anti-human CD120b [3G7A02]; Isotype: Rat IgG2a, κ; Reactivity: Human; Apps: FC; Size: 100 tests
Catalog No. 50-850-777