Learn More
Biolegend PE/Cyanine7 anti-human CD16
Supplier:  Biolegend 360708
Encompass_Preferred
PE/Cyanine7 anti-human CD16 [B73.1]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 100 tests
Catalog No. 50-604-666