Learn More
Biolegend PE/Cyanine7 anti-human CD19
Supplier:  Biolegend 302215
Encompass_Preferred
PE/Cyanine7 anti-human CD19 [HIB19]; Isotype: Mouse IgG1, κ; Reactivity: Human, Chimpanzee, Rhesus; Apps: FC; Size: 25 tests
Catalog No. NC1124548