Learn More
Biolegend PE/Cyanine7 anti-human CD1c
Supplier:  Biolegend 331516
Encompass_Preferred
PE/Cyanine7 anti-human CD1c [L161]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green, Baboon, Cynomolgus, Rhesus; Apps: FC; Size: 100 tests
Catalog No. NC9013385