Learn More
Biolegend PE/Cyanine7 anti-human CD3
Supplier:  Biolegend 300419
Encompass_Preferred
PE/Cyanine7 anti-human CD3 [UCHT1]; Isotype: Mouse IgG1, κ; Reactivity: Human, Cross-Reactivity: Chimpanzee; Apps: FC; Size: 25 tests
Catalog No. 50-165-568