Learn More
Biolegend PE/Cyanine7 anti-human CD4
Supplier:  Biolegend 300512
Encompass_Preferred
PE/Cyanine7 anti-human CD4 [RPA-T4]; Isotype: Mouse IgG1, κ; Reactivity: Human, Chimpanzee; Apps: FC; Size: 100 tests
Catalog No. 50-165-587