Learn More
Biolegend PE/Cyanine7 anti-human CD56 (NCAM)
Supplier:  Biolegend 318317
Encompass_Preferred
PE/Cyanine7 anti-human CD56 (NCAM) [HCD56]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green, Baboon, Cynomolgus, Rhesus; Apps: FC; Size: 25 tests
Catalog No. 50-166-967