Learn More
Biolegend PE/Cyanine7 anti-human CD68
Supplier:  Biolegend 333815
Encompass_Preferred
PE/Cyanine7 anti-human CD68 [Y1/82A]; Isotype: Mouse IgG2b, κ; Reactivity: Human; Apps: ICFC; Size: 25 tests
Catalog No. 50-604-240