Learn More
Biolegend PE/Cyanine7 anti-human CD7
Supplier:  Biolegend 343114
Encompass_Preferred
PE/Cyanine7 anti-human CD7 [CD7-6B7]; Isotype: Mouse IgG2a, κ; Reactivity: Human; Apps: FC; Size: 100 tests
Catalog No. 50-850-694