Learn More
Biolegend PE/Cyanine7 anti-human CD80
Supplier:  Biolegend 305217
Encompass_Preferred
PE/Cyanine7 anti-human CD80 [2D10]; Isotype: Mouse IgG1, κ; Reactivity: Human, Cross-Reactivity: Rhesus; Apps: FC; Size: 25 tests
Catalog No. 50-166-230