Learn More
Biolegend PE/Cyanine7 anti-human CD83
Supplier:  Biolegend 305326
Encompass_Preferred
PE/Cyanine7 anti-human CD83 [HB15e]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green, Baboon, Pigtailed Macaque, Rhesus; Apps: FC; Size: 100 tests
Catalog No. 50-604-051